Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600007 MIM: 180471 | ||||||||||||||||||||
Literature Links: |
FLT3LG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLT3LG - fms related tyrosine kinase 3 ligand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204502.1 | Intron | NP_001191431.1 | ||||
NM_001204503.1 | Intron | NP_001191432.1 | ||||
NM_001278637.1 | Intron | NP_001265566.1 | ||||
NM_001278638.1 | Intron | NP_001265567.1 | ||||
NM_001459.3 | Intron | NP_001450.2 | ||||
XM_005258680.4 | Intron | XP_005258737.3 | ||||
XM_005258681.4 | Intron | XP_005258738.3 | ||||
XM_005258682.4 | Intron | XP_005258739.3 | ||||
XM_005258683.4 | Intron | XP_005258740.3 | ||||
XM_006723116.3 | Intron | XP_006723179.2 | ||||
XM_011526675.2 | Intron | XP_011524977.1 | ||||
XM_011526676.2 | Intron | XP_011524978.1 | ||||
XM_011526677.2 | Intron | XP_011524979.1 | ||||
XM_011526678.2 | Intron | XP_011524980.1 | ||||
XM_011526680.2 | Intron | XP_011524982.1 | ||||
XM_011526682.2 | Intron | XP_011524984.1 | ||||
XM_017026532.1 | Intron | XP_016882021.1 | ||||
XM_017026533.1 | Intron | XP_016882022.1 | ||||
XM_017026534.1 | Intron | XP_016882023.1 | ||||
XM_017026535.1 | Intron | XP_016882024.1 |
RPL13A - ribosomal protein L13a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270491.1 | Intron | NP_001257420.1 | ||||
NM_012423.3 | Intron | NP_036555.1 |
RPS11 - ribosomal protein S11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD32A - small nucleolar RNA, C/D box 32A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD33 - small nucleolar RNA, C/D box 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD34 - small nucleolar RNA, C/D box 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD35A - small nucleolar RNA, C/D box 35A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |