Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTCTTTTGTTTTGTTTTGAGAC[A/G]GAGTTTCACTCTTGTTGCCCAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613910 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF480 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF480 - zinc finger protein 480 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297624.1 | Intron | NP_001284553.1 | ||||
NM_001297625.1 | Intron | NP_001284554.1 | ||||
NM_144684.3 | Intron | NP_653285.2 | ||||
XM_011526465.2 | Intron | XP_011524767.1 |
ZNF766 - zinc finger protein 766 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |