Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGCCTTCTCCTGCTGCCGGCTG[A/T]GCTGGAACAGGTCGGCCAGGAACTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 116897 | ||||||||||||||||||||
Literature Links: |
CEBPA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEBPA - CCAAT/enhancer binding protein alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285829.1 | 401 | UTR 5 | NP_001272758.1 | |||
NM_001287424.1 | 401 | Missense Mutation | CAC,CTC | H,L 119 | NP_001274353.1 | |
NM_001287435.1 | 401 | Missense Mutation | CAC,CTC | H,L 70 | NP_001274364.1 | |
NM_004364.4 | 401 | Missense Mutation | CAC,CTC | H,L 84 | NP_004355.2 |
CEBPA-AS1 - CEBPA antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |