Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGGAATCCGAGGAATGAATGAAT[C/G]AATGAATGAATGAATAAACGAACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607990 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARRDC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARRDC5 - arrestin domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080523.1 | 158 | Intron | NP_001073992.1 | |||
XM_005259616.2 | 158 | Intron | XP_005259673.1 | |||
XM_011528181.2 | 158 | Intron | XP_011526483.1 | |||
XM_017027144.1 | 158 | Intron | XP_016882633.1 | |||
XM_017027145.1 | 158 | Intron | XP_016882634.1 |
UHRF1 - ubiquitin like with PHD and ring finger domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001048201.2 | 158 | Intron | NP_001041666.1 | |||
NM_001290050.1 | 158 | Intron | NP_001276979.1 | |||
NM_001290051.1 | 158 | Intron | NP_001276980.1 | |||
NM_001290052.1 | 158 | Intron | NP_001276981.1 | |||
NM_013282.4 | 158 | UTR 5 | NP_037414.3 | |||
XM_011527942.2 | 158 | Intron | XP_011526244.1 |