Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTGAAAGCAGGAAGGAAGAAAT[A/G]CGCAGGCTCCAGCCGCGTCCCCGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109480 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BSG PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BSG - basigin (Ok blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322243.1 | Intron | NP_001309172.1 | ||||
NM_001728.3 | Intron | NP_001719.2 | ||||
NM_198589.2 | Intron | NP_940991.1 | ||||
NM_198590.2 | Intron | NP_940992.1 | ||||
NM_198591.2 | Intron | NP_940993.1 | ||||
XM_017027173.1 | Intron | XP_016882662.1 |
Set Membership: |
HapMap |