Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGAAGGCACCCATTTACAGCAA[C/T]GTAGGCAGGCTTGCAGACTTTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603984 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF737 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF737 - zinc finger protein 737 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159293.1 | Intron | NP_001152765.1 | ||||
XM_005259697.3 | Intron | XP_005259754.1 | ||||
XM_006722597.3 | Intron | XP_006722660.1 | ||||
XM_011527598.2 | Intron | XP_011525900.1 | ||||
XM_011527599.2 | Intron | XP_011525901.1 | ||||
XM_017026116.1 | Intron | XP_016881605.1 | ||||
XM_017026117.1 | Intron | XP_016881606.1 |