Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAAGCATTCCTTTTACAACCTTA[C/T]CAGCATCTGTGATTTTTTTTAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606957 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF257 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF257 - zinc finger protein 257 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316996.1 | Intron | NP_001303925.1 | ||||
NM_001316997.1 | Intron | NP_001303926.1 | ||||
NM_001316998.1 | Intron | NP_001303927.1 | ||||
NM_033468.3 | Intron | NP_258429.2 |