Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGACCCCGCGACTTCTGGCAGCCC[C/G]TGATCTGCCACGCGCGCCTCAATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147840 MIM: 614088 MIM: 601852 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ICAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ICAM1 - intercellular adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ICAM4 - intercellular adhesion molecule 4 (Landsteiner-Wiener blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039132.2 | 668 | Missense Mutation | CCT,CGT | P,R 182 | NP_001034221.1 | |
NM_001544.4 | 668 | Missense Mutation | CTG,GTG | L,V 208 | NP_001535.1 | |
NM_022377.3 | 668 | Missense Mutation | CTG,GTG | L,V 208 | NP_071772.1 |
ICAM5 - intercellular adhesion molecule 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |