Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTGCCCGTGGGTGCCCGCGGAG[A/G]AAGTGGCATCCGGGCCTGGGCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607573 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP1S PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP1S - microtubule associated protein 1S | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308363.1 | Intron | NP_001295292.1 | ||||
NM_018174.5 | Intron | NP_060644.4 | ||||
XM_017026930.1 | Intron | XP_016882419.1 | ||||
XM_017026931.1 | Intron | XP_016882420.1 |