Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCACGCTCCTGGCGCCAGGTACG[A/G]ATGGAGGACCCCTGCTCCAACACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602982 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARRDC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARRDC2 - arrestin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025604.2 | Intron | NP_001020775.1 | ||||
NM_001286826.1 | Intron | NP_001273755.1 | ||||
NM_015683.1 | Intron | NP_056498.1 |
KCNN1 - potassium calcium-activated channel subfamily N member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |