Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAAATGGAAAGCCACTCCCATGG[A/G]AAGGGAGGGGGTAGCAGCTGGGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606564 | ||||||||||||||||||||
Literature Links: |
PMEPA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PMEPA1 - prostate transmembrane protein, androgen induced 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001255976.1 | 2997 | UTR 3 | NP_001242905.1 | |||
NM_020182.4 | 2997 | UTR 3 | NP_064567.2 | |||
NM_199169.2 | 2997 | UTR 3 | NP_954638.1 | |||
NM_199170.2 | 2997 | UTR 3 | NP_954639.1 | |||
NM_199171.2 | 2997 | UTR 3 | NP_954640.1 |