Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTGTGTATTTCCTGTCTGATGT[A/G]GTTTTTTCCAAGGGAAGTCAGACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172420 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLCG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLCG1 - phospholipase C gamma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002660.2 | Intron | NP_002651.2 | ||||
NM_182811.1 | Intron | NP_877963.1 | ||||
XM_005260438.2 | Intron | XP_005260495.1 | ||||
XM_011528865.1 | Intron | XP_011527167.1 | ||||
XM_011528867.2 | Intron | XP_011527169.1 |
PLCG1-AS1 - PLCG1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |