Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACACACCCATACCTGTGCTTCTG[C/T]AGCAGGGCCACCAGGATGCTCTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 139190 | ||||||||||||||||||||
Literature Links: |
GHRH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GHRH - growth hormone releasing hormone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184731.2 | 316 | Silent Mutation | CTA,CTG | L,L 99 | NP_001171660.1 | |
NM_021081.5 | 316 | Silent Mutation | CTA,CTG | L,L 99 | NP_066567.1 | |
XM_011528784.2 | 316 | Silent Mutation | CTA,CTG | L,L 99 | XP_011527086.1 | |
XM_011528788.2 | 316 | Silent Mutation | CTA,CTG | L,L 99 | XP_011527090.1 |