Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGATTTGCTTTTAGATACTTTT[C/T]TGCTGTGAGAATGTAAGATGGATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604803 | ||||||||||||||||||||
Literature Links: |
CBR3-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CBR3-AS1 - CBR3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DOPEY2 - dopey family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320714.1 | 171 | UTR 5 | NP_001307643.1 | |||
NM_005128.3 | 171 | UTR 5 | NP_005119.2 | |||
XM_017028509.1 | 171 | Intron | XP_016883998.1 |