Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGCAGGTCTGGCCCATGGAAGG[C/G]AGGGGAGGGGGCCCCGACGGGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COMT - catechol-O-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000754.3 | Intron | NP_000745.1 | ||||
NM_001135161.1 | Intron | NP_001128633.1 | ||||
NM_001135162.1 | Intron | NP_001128634.1 | ||||
NM_007310.2 | Intron | NP_009294.1 | ||||
XM_011529886.1 | Intron | XP_011528188.1 | ||||
XM_017028594.1 | Intron | XP_016884083.1 | ||||
XM_017028595.1 | Intron | XP_016884084.1 |
MIR4761 - microRNA 4761 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |