Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCGCGGAACCTTAGGCCTGGCC[A/C]TGGTCTCCGAGCGCGGGTTCGCCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194548 | ||||||||||||||||||||
Literature Links: |
USP41 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USP41 - ubiquitin specific peptidase 41 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF74 - zinc finger protein 74 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256523.1 | 204 | UTR 5 | NP_001243452.1 | |||
NM_001256524.1 | 204 | UTR 5 | NP_001243453.1 | |||
NM_001256525.1 | 204 | UTR 5 | NP_001243454.1 | |||
NM_003426.3 | 204 | UTR 5 | NP_003417.2 |