Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TAAGGACCAGAGTCCCTGTGTAACA[A/C]AGCTAGTTAATGAAAGATGTGGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608222 MIM: 610440 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADSL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADSL - adenylosuccinate lyase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000026.3 | Intron | NP_000017.1 | ||||
NM_001123378.2 | Intron | NP_001116850.1 | ||||
NM_001317923.1 | Intron | NP_001304852.1 | ||||
XM_011529977.2 | Intron | XP_011528279.1 | ||||
XM_011529979.1 | Intron | XP_011528281.1 | ||||
XM_011529980.2 | Intron | XP_011528282.1 | ||||
XM_017028636.1 | Intron | XP_016884125.1 | ||||
XM_017028637.1 | Intron | XP_016884126.1 | ||||
XM_017028638.1 | Intron | XP_016884127.1 | ||||
XM_017028639.1 | Intron | XP_016884128.1 | ||||
XM_017028640.1 | Intron | XP_016884129.1 |
SGSM3 - small G protein signaling modulator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301849.1 | Intron | NP_001288778.1 | ||||
NM_015705.5 | Intron | NP_056520.2 | ||||
XM_005261572.2 | Intron | XP_005261629.1 | ||||
XM_005261573.3 | Intron | XP_005261630.1 | ||||
XM_005261574.4 | Intron | XP_005261631.1 | ||||
XM_005261575.1 | Intron | XP_005261632.1 | ||||
XM_011530158.1 | Intron | XP_011528460.1 | ||||
XM_011530159.1 | Intron | XP_011528461.1 | ||||
XM_011530160.1 | Intron | XP_011528462.1 | ||||
XM_011530161.1 | Intron | XP_011528463.1 | ||||
XM_011530162.2 | Intron | XP_011528464.1 | ||||
XM_017028775.1 | Intron | XP_016884264.1 | ||||
XM_017028776.1 | Intron | XP_016884265.1 | ||||
XM_017028777.1 | Intron | XP_016884266.1 | ||||
XM_017028778.1 | Intron | XP_016884267.1 | ||||
XM_017028779.1 | Intron | XP_016884268.1 | ||||
XM_017028780.1 | Intron | XP_016884269.1 |