Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTGGTCAGGACCAGCCTCTCCC[A/C]GGGAGAGGCATTTGAACTGAGGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 188595 | ||||||||||||||||||||
Literature Links: |
LOC100996598 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100996598 - uncharacterized LOC100996598 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEF - TEF, PAR bZIP transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145398.2 | Intron | NP_001138870.1 | ||||
NM_003216.3 | Intron | NP_003207.1 |