Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACGTAGAGGGCGTTGATTGGGA[A/C]CGAGTACAGGGCCCGCGCATGCGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614637 MIM: 152690 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DESI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DESI1 - desumoylating isopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015704.2 | Intron | NP_056519.1 | ||||
XM_005261571.4 | Intron | XP_005261628.1 |
XRCC6 - X-ray repair cross complementing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288976.1 | Intron | NP_001275905.1 | ||||
NM_001288977.1 | Intron | NP_001275906.1 | ||||
NM_001288978.1 | Intron | NP_001275907.1 | ||||
NM_001469.4 | Intron | NP_001460.1 |