Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTTCTCGAAGTCCAGCCTTTTA[G/T]GACAGTTTCACAGCTGCCTCTAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603107 | ||||||||||||||||||||
Literature Links: |
TCF20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCF20 - transcription factor 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005650.3 | Intron | NP_005641.1 | ||||
NM_181492.2 | Intron | NP_852469.1 | ||||
XM_005261722.3 | Intron | XP_005261779.1 | ||||
XM_006724313.3 | Intron | XP_006724376.1 | ||||
XM_011530353.2 | Intron | XP_011528655.1 | ||||
XM_011530354.2 | Intron | XP_011528656.1 |