Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCCAGCTCTTGCATGACTTGCT[A/G]TGTGACCCCGGGCAAAGTCCTTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615903 MIM: 185261 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C22orf15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C22orf15 - chromosome 22 open reading frame 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CHCHD10 - coiled-coil-helix-coiled-coil-helix domain containing 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMP11 - matrix metallopeptidase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005940.4 | Intron | NP_005931.2 |