Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGGCGGGACCACAGTGGTGTCC[A/G]AGAAGTCAGGCACGTAGCTCAGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 153620 MIM: 610367 | ||||||||||||||||||||
Literature Links: |
MIF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIF - macrophage migration inhibitory factor (glycosylation-inhibiting factor) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002415.1 | Intron | NP_002406.1 |
MIF-AS1 - MIF antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC2A11 - solute carrier family 2 member 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |