Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTTTGTTCTTGATATTAATCTC[C/T]GAAATTAATGGACATGCCTCTGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604251 | ||||||||||||||||||||
Literature Links: |
CABIN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CABIN1 - calcineurin binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199281.1 | Intron | NP_001186210.1 | ||||
NM_001201429.1 | Intron | NP_001188358.1 | ||||
NM_012295.3 | Intron | NP_036427.1 | ||||
XM_005261415.2 | Intron | XP_005261472.1 | ||||
XM_005261416.2 | Intron | XP_005261473.1 | ||||
XM_005261417.2 | Intron | XP_005261474.1 | ||||
XM_005261419.2 | Intron | XP_005261476.1 | ||||
XM_006724179.1 | Intron | XP_006724242.1 | ||||
XM_006724180.1 | Intron | XP_006724243.1 | ||||
XM_006724181.1 | Intron | XP_006724244.1 | ||||
XM_011530026.1 | Intron | XP_011528328.1 | ||||
XM_011530029.2 | Intron | XP_011528331.1 | ||||
XM_011530030.2 | Intron | XP_011528332.1 | ||||
XM_017028680.1 | Intron | XP_016884169.1 | ||||
XM_017028681.1 | Intron | XP_016884170.1 | ||||
XM_017028682.1 | Intron | XP_016884171.1 | ||||
XM_017028683.1 | Intron | XP_016884172.1 | ||||
XM_017028684.1 | Intron | XP_016884173.1 |