Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTGATTGCAGGCAGTAGAGTCA[A/G]GTGATAGGAGGCAGAGTGGCTGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123620 MIM: 123630 | ||||||||||||||||||||
Literature Links: |
CRYBB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRYBB2 - crystallin beta B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000496.2 | 208 | Intron | NP_000487.1 | |||
XM_006724141.3 | 208 | UTR 5 | XP_006724204.1 | |||
XM_011529900.2 | 208 | UTR 5 | XP_011528202.1 |
CRYBB3 - crystallin beta B3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |