Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAGGAGACCCCGTCTCAAAAAC[A/C]AAACAAAACAAACAAATAAAACAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HORMAD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HORMAD2 - HORMA domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011529914.2 | Intron | XP_011528216.1 | ||||
XM_011529917.2 | Intron | XP_011528219.1 | ||||
XM_011529918.1 | Intron | XP_011528220.1 | ||||
XM_017028621.1 | Intron | XP_016884110.1 | ||||
XM_017028622.1 | Intron | XP_016884111.1 | ||||
XM_017028624.1 | Intron | XP_016884113.1 | ||||
XM_017028625.1 | Intron | XP_016884114.1 | ||||
XM_017028626.1 | Intron | XP_016884115.1 |
HORMAD2-AS1 - HORMAD2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |