Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGTCTTTTCACCACTGATTTGC[C/T]TTTGCACATTTGTCAAAATTAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300117 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM133A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
FAM133A - family with sequence similarity 133 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171109.1 | Intron | NP_001164580.1 | ||||
NM_001171110.1 | Intron | NP_001164581.1 | ||||
NM_001171111.1 | Intron | NP_001164582.1 | ||||
NM_173698.2 | Intron | NP_775969.1 | ||||
XM_006724641.3 | Intron | XP_006724704.1 | ||||
XM_011530925.2 | Intron | XP_011529227.1 | ||||
XM_011530926.2 | Intron | XP_011529228.1 | ||||
XM_011530927.2 | Intron | XP_011529229.1 | ||||
XM_011530928.1 | Intron | XP_011529230.1 | ||||
XM_011530929.1 | Intron | XP_011529231.1 | ||||
XM_011530931.2 | Intron | XP_011529233.1 | ||||
XM_017029454.1 | Intron | XP_016884943.1 | ||||
XM_017029455.1 | Intron | XP_016884944.1 | ||||
XM_017029456.1 | Intron | XP_016884945.1 |
NAP1L3 - nucleosome assembly protein 1 like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |