Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCAACCTAGATGCCCATCAACGG[C/T]GGGCTAAAGAAAATGTACATATATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300255 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OGT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OGT - O-linked N-acetylglucosamine (GlcNAc) transferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181672.2 | Intron | NP_858058.1 | ||||
NM_181673.2 | Intron | NP_858059.1 | ||||
XM_017029907.1 | Intron | XP_016885396.1 | ||||
XM_017029908.1 | Intron | XP_016885397.1 |