Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGCCGGCCGCGGGCCGGGTGGGG[A/T]TGCCTCTGCACGTGAAGTGGCCGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300081 MIM: 312173 MIM: 300394 | ||||||||||||||||||||
Literature Links: |
DNASE1L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNASE1L1 - deoxyribonuclease I-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009932.2 | 327 | Intron | NP_001009932.1 | |||
NM_001009933.2 | 327 | Intron | NP_001009933.1 | |||
NM_001009934.2 | 327 | Intron | NP_001009934.1 | |||
NM_001303620.1 | 327 | Intron | NP_001290549.1 | |||
NM_006730.3 | 327 | Intron | NP_006721.1 | |||
XM_005277829.4 | 327 | Intron | XP_005277886.1 | |||
XM_011531122.2 | 327 | Intron | XP_011529424.1 | |||
XM_017029332.1 | 327 | Intron | XP_016884821.1 |
RPL10 - ribosomal protein L10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAZ - tafazzin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000116.4 | 327 | Missense Mutation | ATG,TTG | M,L 1 | NP_000107.1 | |
NM_001303465.1 | 327 | Missense Mutation | ATG,TTG | M,L 1 | NP_001290394.1 | |
NM_181311.3 | 327 | Missense Mutation | ATG,TTG | M,L 1 | NP_851828.1 | |
NM_181312.3 | 327 | Missense Mutation | ATG,TTG | M,L 1 | NP_851829.1 | |
NM_181313.3 | 327 | Missense Mutation | ATG,TTG | M,L 1 | NP_851830.1 | |
XM_006724836.1 | 327 | Missense Mutation | ATG,TTG | M,L 1 | XP_006724899.1 | |
XM_006724837.1 | 327 | Missense Mutation | ATG,TTG | M,L 1 | XP_006724900.1 | |
XM_006724839.1 | 327 | Missense Mutation | ATG,TTG | M,L 1 | XP_006724902.1 | |
XM_006724841.3 | 327 | Intron | XP_006724904.1 | |||
XM_006724842.3 | 327 | Intron | XP_006724905.1 | |||
XM_011531189.1 | 327 | Missense Mutation | ATG,TTG | M,L 1 | XP_011529491.1 | |
XM_011531191.2 | 327 | Intron | XP_011529493.1 | |||
XM_017029761.1 | 327 | Missense Mutation | ATG,TTG | M,L 1 | XP_016885250.1 | |
XM_017029762.1 | 327 | Missense Mutation | ATG,TTG | M,L 1 | XP_016885251.1 | |
XM_017029763.1 | 327 | Missense Mutation | ATG,TTG | M,L 1 | XP_016885252.1 | |
XM_017029764.1 | 327 | Intron | XP_016885253.1 | |||
XM_017029765.1 | 327 | UTR 5 | XP_016885254.1 |