Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGGAAAACAATACAGTGGGTCC[C/T]CAAAAAATTAAAAATAGAATTACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC107985657 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC107985657 - uncharacterized LOC107985657 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCEANC - transcription elongation factor A N-terminal and central domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297563.1 | Intron | NP_001284492.1 | ||||
NM_001297564.1 | Intron | NP_001284493.1 | ||||
NM_152634.3 | Intron | NP_689847.2 | ||||
XM_017029316.1 | Intron | XP_016884805.1 | ||||
XM_017029317.1 | Intron | XP_016884806.1 | ||||
XM_017029318.1 | Intron | XP_016884807.1 | ||||
XM_017029319.1 | Intron | XP_016884808.1 |