Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGTGGCGCTCATCGAGGGACGCC[C/T]GGCCCAAGTGGTAAGTTGAAAAATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300079 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
XIAP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
XIAP - X-linked inhibitor of apoptosis | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167.3 | 70 | Intron | NP_001158.2 | |||
NM_001204401.1 | 70 | UTR 5 | NP_001191330.1 | |||
XM_006724754.2 | 70 | UTR 5 | XP_006724817.1 | |||
XM_011531329.2 | 70 | Intron | XP_011529631.1 |
Set Membership: |
HapMap |