Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCTGTTTGCACTCATCATGTC[C/T]GTTTACTTGATTGAGATGCTTCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300079 | ||||||||||||||||||||
Literature Links: |
XIAP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
XIAP - X-linked inhibitor of apoptosis | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167.3 | Intron | NP_001158.2 | ||||
NM_001204401.1 | Intron | NP_001191330.1 | ||||
XM_006724754.2 | Intron | XP_006724817.1 | ||||
XM_011531329.2 | Intron | XP_011529631.1 |