Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCTGATCCAATGGCATTATTGG[A/G]AATTTACATATGCTTTAGTATCATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608151 | ||||||||||||||||||||
Literature Links: |
WDR19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDR19 - WD repeat domain 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317924.1 | Intron | NP_001304853.1 | ||||
NM_025132.3 | Intron | NP_079408.3 | ||||
XM_011513724.1 | Intron | XP_011512026.1 | ||||
XM_011513725.2 | Intron | XP_011512027.1 | ||||
XM_011513726.2 | Intron | XP_011512028.1 | ||||
XM_011513727.1 | Intron | XP_011512029.1 | ||||
XM_017008501.1 | Intron | XP_016863990.1 |