Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCACATTATGAAATATGACTTG[C/T]TGACCATGGAACAGTGCTTGGATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMIM14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMIM14 - small integral membrane protein 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317896.1 | 4757 | UTR 3 | NP_001304825.1 | |||
NM_001317897.1 | 4757 | UTR 3 | NP_001304826.1 | |||
NM_174921.2 | 4757 | UTR 3 | NP_777581.1 | |||
XM_011513640.2 | 4757 | UTR 3 | XP_011511942.1 | |||
XM_017007862.1 | 4757 | UTR 3 | XP_016863351.1 | |||
XM_017007863.1 | 4757 | UTR 3 | XP_016863352.1 |
UGDH-AS1 - UGDH antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |