Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCCTCAAGGATTATGTCATGCA[C/T]GCCTGTCAGAATAGGGCTCCTGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602846 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2K PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBE2K - ubiquitin conjugating enzyme E2 K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111112.1 | Intron | NP_001104582.1 | ||||
NM_001111113.1 | Intron | NP_001104583.1 | ||||
NM_001312646.1 | Intron | NP_001299575.1 | ||||
NM_001312647.1 | Intron | NP_001299576.1 | ||||
NM_001312648.1 | Intron | NP_001299577.1 | ||||
NM_005339.4 | Intron | NP_005330.1 |