Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTTTGAGGTAATATGAGCTATT[C/T]GACTAATCCAGATAAGGAATTTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603248 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMPR1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BMPR1B - bone morphogenetic protein receptor type 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001203.2 | Intron | NP_001194.1 | ||||
NM_001256792.1 | Intron | NP_001243721.1 | ||||
NM_001256793.1 | Intron | NP_001243722.1 | ||||
NM_001256794.1 | Intron | NP_001243723.1 | ||||
XM_011532201.2 | Intron | XP_011530503.1 | ||||
XM_017008558.1 | Intron | XP_016864047.1 | ||||
XM_017008559.1 | Intron | XP_016864048.1 | ||||
XM_017008560.1 | Intron | XP_016864049.1 | ||||
XM_017008561.1 | Intron | XP_016864050.1 |
BMPR1B-AS1 - BMPR1B antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |