Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCCAGAGTGAAAAGGACAGTTC[C/T]ATTGAGGGTTAGGCGTGAATGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610151 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
METAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
METAP1 - methionyl aminopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015143.2 | Intron | NP_055958.2 | ||||
XM_011531777.2 | Intron | XP_011530079.1 | ||||
XM_011531778.1 | Intron | XP_011530080.1 | ||||
XM_011531779.1 | Intron | XP_011530081.1 | ||||
XM_017007914.1 | Intron | XP_016863403.1 | ||||
XM_017007915.1 | Intron | XP_016863404.1 | ||||
XM_017007916.1 | Intron | XP_016863405.1 | ||||
XM_017007917.1 | Intron | XP_016863406.1 |
MIR3684 - microRNA 3684 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |