Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAAAACAGACCCTTCTGTCCTCT[A/G]TATTTGTAGTTCTACCCTCCTGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610851 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP1AR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AP1AR - adaptor related protein complex 1 associated regulatory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128426.2 | Intron | NP_001121898.1 | ||||
NM_018569.5 | Intron | NP_061039.3 | ||||
XM_017008378.1 | Intron | XP_016863867.1 | ||||
XM_017008379.1 | Intron | XP_016863868.1 | ||||
XM_017008380.1 | Intron | XP_016863869.1 |
LOC105377371 - uncharacterized LOC105377371 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |