Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGGAGACGGAAATGTCCGAAGGC[C/T]GCAGTACTTGACCCTGTATTTTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612026 | ||||||||||||||||||||
Literature Links: |
LARP7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LARP7 - La ribonucleoprotein domain family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267039.1 | 240 | UTR 5 | NP_001253968.1 | |||
NM_015454.2 | 240 | Intron | NP_056269.1 | |||
NM_016648.3 | 240 | UTR 5 | NP_057732.2 |
ZGRF1 - zinc finger GRF-type containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018392.4 | 240 | Intron | NP_060862.3 | |||
XM_005263115.3 | 240 | Intron | XP_005263172.1 | |||
XM_011532091.2 | 240 | Intron | XP_011530393.1 | |||
XM_011532092.2 | 240 | Intron | XP_011530394.1 | |||
XM_011532093.2 | 240 | Intron | XP_011530395.1 | |||
XM_011532094.2 | 240 | Intron | XP_011530396.1 | |||
XM_011532096.2 | 240 | Intron | XP_011530398.1 | |||
XM_011532097.2 | 240 | Intron | XP_011530399.1 | |||
XM_011532098.2 | 240 | Intron | XP_011530400.1 | |||
XM_011532099.2 | 240 | Intron | XP_011530401.1 | |||
XM_017008369.1 | 240 | Intron | XP_016863858.1 | |||
XM_017008370.1 | 240 | Intron | XP_016863859.1 | |||
XM_017008371.1 | 240 | Intron | XP_016863860.1 | |||
XM_017008372.1 | 240 | Intron | XP_016863861.1 | |||
XM_017008373.1 | 240 | Intron | XP_016863862.1 |