Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGTGCCGTTGTCAGAGTTCGCAA[A/C]TCGAGCGCCCAGAGGGCTCGCGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601291 | ||||||||||||||||||||
Literature Links: |
UGT8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UGT8 - UDP glycosyltransferase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128174.2 | 15 | UTR 5 | NP_001121646.1 | |||
NM_001322112.1 | 15 | UTR 5 | NP_001309041.1 | |||
NM_001322113.1 | 15 | UTR 5 | NP_001309042.1 | |||
NM_001322114.1 | 15 | Intron | NP_001309043.1 | |||
NM_003360.4 | 15 | Intron | NP_003351.2 |