Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGACCTGTTAGTGAACATATGC[A/C]TGCACATATACGGTACATAAATACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611565 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1109 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1109 - KIAA1109 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015312.3 | Intron | NP_056127.2 | ||||
XM_005263282.1 | Intron | XP_005263339.1 | ||||
XM_005263287.1 | Intron | XP_005263344.1 | ||||
XM_006714344.1 | Intron | XP_006714407.1 | ||||
XM_011532319.1 | Intron | XP_011530621.1 | ||||
XM_011532320.2 | Intron | XP_011530622.1 | ||||
XM_011532321.2 | Intron | XP_011530623.1 | ||||
XM_011532322.1 | Intron | XP_011530624.1 | ||||
XM_011532323.1 | Intron | XP_011530625.1 | ||||
XM_011532324.1 | Intron | XP_011530626.1 | ||||
XM_011532325.1 | Intron | XP_011530627.1 | ||||
XM_011532326.1 | Intron | XP_011530628.1 | ||||
XM_011532327.1 | Intron | XP_011530629.1 | ||||
XM_011532330.1 | Intron | XP_011530632.1 | ||||
XM_017008695.1 | Intron | XP_016864184.1 | ||||
XM_017008696.1 | Intron | XP_016864185.1 | ||||
XM_017008697.1 | Intron | XP_016864186.1 | ||||
XM_017008698.1 | Intron | XP_016864187.1 | ||||
XM_017008699.1 | Intron | XP_016864188.1 |