Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATCAGTATATGTCTCTCTTGCA[C/T]ACACACATTCACACCATGTTTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105377623 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105377623 - uncharacterized LOC105377623 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP38 - ubiquitin specific peptidase 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290325.1 | Intron | NP_001277254.1 | ||||
NM_001290326.1 | Intron | NP_001277255.1 | ||||
NM_032557.6 | Intron | NP_115946.2 | ||||
XM_011532360.2 | Intron | XP_011530662.1 |