Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTTCAGGTTCGTGGGCCTGCAG[A/G]GGAGAGACTCGAACTCGTGGAACCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604439 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GAB1 - GRB2 associated binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002039.3 | 182 | UTR 5 | NP_002030.2 | |||
NM_207123.2 | 182 | UTR 5 | NP_997006.1 | |||
XM_006714167.3 | 182 | Intron | XP_006714230.1 | |||
XM_006714168.3 | 182 | Intron | XP_006714231.1 | |||
XM_017007966.1 | 182 | Intron | XP_016863455.1 | |||
XM_017007967.1 | 182 | Intron | XP_016863456.1 | |||
XM_017007968.1 | 182 | Intron | XP_016863457.1 | |||
XM_017007969.1 | 182 | Intron | XP_016863458.1 | |||
XM_017007970.1 | 182 | Intron | XP_016863459.1 | |||
XM_017007971.1 | 182 | Intron | XP_016863460.1 |
MIR3139 - microRNA 3139 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |