Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTATTTTGAAAACAGCTTTATAC[A/G]TACTTCTTTAAGAATACTGCACGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604439 | ||||||||||||||||||||
Literature Links: |
GAB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GAB1 - GRB2 associated binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002039.3 | Intron | NP_002030.2 | ||||
NM_207123.2 | Intron | NP_997006.1 | ||||
XM_006714167.3 | Intron | XP_006714230.1 | ||||
XM_006714168.3 | Intron | XP_006714231.1 | ||||
XM_017007966.1 | Intron | XP_016863455.1 | ||||
XM_017007967.1 | Intron | XP_016863456.1 | ||||
XM_017007968.1 | Intron | XP_016863457.1 | ||||
XM_017007969.1 | Intron | XP_016863458.1 | ||||
XM_017007970.1 | Intron | XP_016863459.1 | ||||
XM_017007971.1 | Intron | XP_016863460.1 |
MIR3139 - microRNA 3139 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |