Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAAGAATGTGTACTTTCTCTCT[C/T]TTTTTTTTAACACTCACATGCTTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608674 | ||||||||||||||||||||
Literature Links: |
SH3D19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SH3D19 - SH3 domain containing 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009555.3 | Intron | NP_001009555.3 | ||||
NM_001128923.1 | Intron | NP_001122395.1 | ||||
NM_001128924.1 | Intron | NP_001122396.1 | ||||
NM_001243349.1 | Intron | NP_001230278.1 | ||||
XM_005262767.2 | Intron | XP_005262824.1 | ||||
XM_011531646.1 | Intron | XP_011529948.1 | ||||
XM_011531647.1 | Intron | XP_011529949.1 | ||||
XM_011531648.1 | Intron | XP_011529950.1 | ||||
XM_011531649.1 | Intron | XP_011529951.1 | ||||
XM_017007778.1 | Intron | XP_016863267.1 | ||||
XM_017007779.1 | Intron | XP_016863268.1 | ||||
XM_017007780.1 | Intron | XP_016863269.1 | ||||
XM_017007781.1 | Intron | XP_016863270.1 | ||||
XM_017007782.1 | Intron | XP_016863271.1 | ||||
XM_017007783.1 | Intron | XP_016863272.1 |