Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATTTACATTCCCCAGAATGTGCC[C/T]CTAGTGTGAATTTTGTATTCTTATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605928 | ||||||||||||||||||||
Literature Links: |
ARFIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARFIP1 - ADP ribosylation factor interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025593.2 | 297 | UTR 3 | NP_001020764.1 | |||
NM_001025595.2 | 297 | UTR 3 | NP_001020766.1 | |||
NM_001287431.1 | 297 | UTR 3 | NP_001274360.1 | |||
NM_001287432.1 | 297 | UTR 3 | NP_001274361.1 | |||
NM_001287433.1 | 297 | UTR 3 | NP_001274362.1 | |||
NM_014447.3 | 297 | UTR 3 | NP_055262.1 | |||
XM_011531868.2 | 297 | UTR 3 | XP_011530170.1 | |||
XM_011531869.2 | 297 | UTR 3 | XP_011530171.1 | |||
XM_011531870.2 | 297 | UTR 3 | XP_011530172.1 | |||
XM_011531871.2 | 297 | UTR 3 | XP_011530173.1 | |||
XM_011531872.1 | 297 | UTR 3 | XP_011530174.1 |
FHDC1 - FH2 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033393.2 | 297 | Intron | NP_203751.2 | |||
XM_005263319.3 | 297 | Intron | XP_005263376.1 | |||
XM_006714393.3 | 297 | Intron | XP_006714456.1 | |||
XM_011532388.1 | 297 | UTR 5 | XP_011530690.1 | |||
XM_011532389.1 | 297 | Intron | XP_011530691.1 | |||
XM_017008787.1 | 297 | Intron | XP_016864276.1 |