Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTCTATGGTTCTGTGTAGTCAC[A/T]AAACAGTCTAATTTTAGTTATAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162642 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPY2R PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NPY2R - neuropeptide Y receptor Y2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000910.3 | Intron | NP_000901.1 | ||||
XM_005263033.4 | Intron | XP_005263090.1 | ||||
XM_005263034.4 | Intron | XP_005263091.1 |
Set Membership: |
HapMap |