Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTCCCTGGGCAGGTGGCCAACT[A/G]GAACAGCAGGCTCTAAGTCCCATGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616210 MIM: 231675 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf46 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C4orf46 - chromosome 4 open reading frame 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008393.3 | Intron | NP_001008394.1 |
ETFDH - electron transfer flavoprotein dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281737.1 | Intron | NP_001268666.1 | ||||
NM_001281738.1 | Intron | NP_001268667.1 | ||||
NM_004453.3 | Intron | NP_004444.2 |
Set Membership: |
HapMap |