Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCCAGCGGGCAGGGTGACTTCC[C/T]TTTTCTGCCCACTCTGGTAACTTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607245 MIM: 609683 | ||||||||||||||||||||
Literature Links: |
AP4B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AP4B1 - adaptor related protein complex 4 beta 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253852.2 | 126 | Intron | NP_001240781.1 | |||
NM_001253853.2 | 126 | Intron | NP_001240782.1 | |||
NM_001308312.1 | 126 | Intron | NP_001295241.1 | |||
NM_006594.4 | 126 | Intron | NP_006585.2 | |||
XM_011540523.2 | 126 | Intron | XP_011538825.1 | |||
XM_011540524.2 | 126 | Intron | XP_011538826.1 | |||
XM_011540525.2 | 126 | Intron | XP_011538827.1 | |||
XM_011540528.2 | 126 | Intron | XP_011538830.1 | |||
XM_017000088.1 | 126 | Intron | XP_016855577.1 | |||
XM_017000089.1 | 126 | Intron | XP_016855578.1 | |||
XM_017000090.1 | 126 | Intron | XP_016855579.1 | |||
XM_017000091.1 | 126 | Intron | XP_016855580.1 | |||
XM_017000092.1 | 126 | Intron | XP_016855581.1 | |||
XM_017000093.1 | 126 | Intron | XP_016855582.1 |
AP4B1-AS1 - AP4B1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DCLRE1B - DNA cross-link repair 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319946.1 | 126 | UTR 5 | NP_001306875.1 | |||
NM_001319947.1 | 126 | Intron | NP_001306876.1 | |||
NM_022836.3 | 126 | UTR 5 | NP_073747.1 |