Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCTTCAACCGCCACACAGTCAC[A/C]TTATCAAATAGTGTGTTCACCTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 613890 | ||||||||||||||||||||
Literature Links: |
HSD3B2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSD3B2 - hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000198.3 | 1209 | Silent Mutation | ACA,ACC | T,T 320 | NP_000189.1 | |
NM_001166120.1 | 1209 | Silent Mutation | ACA,ACC | T,T 320 | NP_001159592.1 |