Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTCTGGTGTGGGAATGGGGGCA[A/G]GGGATGCTAAGCCAAGCCAGCTCCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609957 | |||||||||||||||||||||||
Literature Links: |
FAM19A3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FAM19A3 - family with sequence similarity 19 member A3, C-C motif chemokine like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004440.1 | Intron | NP_001004440.1 | ||||
NM_182759.2 | Intron | NP_877436.1 | ||||
XM_005270766.3 | Intron | XP_005270823.1 | ||||
XM_006710582.3 | Intron | XP_006710645.1 |
LOC105378910 - uncharacterized LOC105378910 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPM1J - protein phosphatase, Mg2+/Mn2+ dependent 1J | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |